Assay Details
Target Gene Details
Entrez Gene ID: | 67920 |
Gene Name: | MAK16 homolog (S. cerevisiae) |
Gene Aliases: |
2600016B03Rik, AI314911, Rbm13 |
Location: |
Chr.8:31159468-31168724 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mak16 | NM_026453.3 | 10 | 932 | NP_080729.1 |
| AK015369.1 | 10 | 973 | BAB29815.1 | |
| AK075982.1 | 10 | 933 | BAC36094.1 | |
| AK075983.1 | 10 | 933 | BAC36095.1 | |
| BC013079.1 | 10 | 929 | AAH13079.1 |
Target Gene Details
Entrez Gene ID: | 234138 |
Gene Name: | TELO2 interacting protein 2 |
Gene Aliases: |
- |
Location: |
Chr.8:31150316-31164703 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tti2 | NM_001199988.1 | NP_001186917.1 | ||
| NR_103719.1 | ||||
| AK045392.1 | 7 | 2476 | BAC32341.1 | |
| AK084242.1 | BAC39146.1 | |||
| AK139425.1 | BAE24007.1 |
More Information
Set Membership: |
|

Genomic Map