Assay Details
Target Gene Details
Entrez Gene ID: | 212528 |
Gene Name: | tRNA methyltransferase 1 |
Gene Aliases: |
6720406L13Rik, 6720477L24, D8Ertd812e |
Location: |
Chr.8:84688725-84699808 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt1 | NM_001164559.1 | 2 | 333 | NP_001158031.1 |
| NM_001164560.1 | 2 | 257 | NP_001158032.1 | |
| NM_198020.2 | 2 | 360 | NP_932137.2 | |
| AK032945.1 | 2 | 268 | ||
| AK040715.1 | 2 | 222 | ||
| AK078395.1 | 2 | 298 | ||
| AK084775.1 | 2 | 298 | ||
| AK139463.1 | 2 | 292 | ||
| AK155251.1 | 2 | 325 | ||
| AK159467.1 | 2 | 298 | BAE35108.1 | |
| AK170412.1 | 2 | 222 | BAE41778.1 | |
| AK171002.1 | 2 | 325 | BAE42173.1 | |
| BC027404.1 | 2 | 329 | AAH27404.1 | |
| CN528783.1 |
More Information
Set Membership: |
|

Genomic Map