Assay Details
Target Gene Details
Entrez Gene ID: | 77781 |
Gene Name: | EPM2A (laforin) interacting protein 1 |
Gene Aliases: |
A930003G21Rik, mKIAA0766 |
Location: |
Chr.9:111271845-111279093 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epm2aip1 | NM_175266.4 | 1 | 1203 | NP_780475.1 |
| AK030084.1 | 1 | 1120 | BAC26774.1 | |
| AK032284.1 | 1 | 1174 | BAC27793.1 | |
| AK043518.1 | 1 | 1161 | ||
| AK122366.1 | 1 | 296 | BAC65648.1 | |
| BC018474.1 | 1 | 1207 | AAH18474.1 |

Genomic Map