Assay Details
Target Gene Details
Entrez Gene ID: | 11733 |
Gene Name: | ankyrin 1, erythroid |
Gene Aliases: |
Ank-1, nb, pale |
Location: |
Chr.8:22974836-23150502 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ank1 | NM_001110783.3 | 1 | 368 | NP_001104253.1 |
| NM_001277280.2 | 1 | 368 | NP_001264209.1 | |
| AK147278.1 | 1 | 323 | BAE27815.1 | |
| AK158131.1 | 1 | 361 | BAE34375.1 | |
| CJ105945.1 | 1 | 368 |

Genomic Map