Assay Details
Target Gene Details
Entrez Gene ID: | 12774 |
Gene Name: | chemokine (C-C motif) receptor 5 |
Gene Aliases: |
AM4-7, CD195, Cmkbr5 |
Location: |
Chr.9:124121543-124127183 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccr5 | NM_009917.5 | 2 | 1028 | NP_034047.2 |
| AK141906.1 | 2 | 999 | BAE24879.1 | |
| AK154595.1 | 2 | 1029 | BAE32698.1 | |
| AK155628.1 | 2 | 1002 | BAE33354.1 | |
| AK155632.1 | 2 | 999 | ||
| AK155867.1 | 2 | 993 | BAE33471.1 | |
| AK170303.1 | 2 | 999 | BAE41700.1 | |
| BC103574.1 | 2 | 967 | AAI03575.1 | |
| BC103586.1 | 2 | 967 | AAI03587.1 | |
| BC103587.1 | 2 | 967 | AAI03588.1 | |
| BC105643.1 | 2 | 968 | ||
| D83648.1 | 2 | 998 | BAA12024.1 | |
| U47036.1 | 2 | 957 | AAC52454.1 | |
| X94151.1 | 2 | 986 | CAA63867.1 |

Genomic Map