Assay Details
Target Gene Details
Entrez Gene ID: | 20516 |
Gene Name: | solute carrier family 20, member 2 |
Gene Aliases: |
MolPit2, Pit-2, Pit2, Ram-1, Ram1 |
Location: |
Chr.8:22476700-22569616 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc20a2 | NM_011394.3 | 2 | 415 | NP_035524.2 |
| AF196476.1 | 1 | 183 | AAG28493.1 | |
| AK087507.1 | 2 | 239 | ||
| AK157206.1 | 2 | 289 | BAE33999.1 | |
| AK170985.1 | 2 | 381 | BAE42159.1 | |
| BC046510.1 | 2 | 337 | AAH46510.1 | |
| BY011867.1 | ||||
| BY237432.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map