Assay Details
Target Gene Details
Entrez Gene ID: | 72254 |
Gene Name: | RIKEN cDNA 1700030K09 gene |
Gene Aliases: |
- |
Location: |
Chr.8:72443880-72460541 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700030K09Rik | NM_028170.2 | 2 | 585 | NP_082446.2 |
| AK006557.1 | 1 | 395 | BAB24653.2 | |
| AK089918.1 | 2 | 527 | ||
| AK133815.1 | 2 | 589 | BAE21858.1 | |
| BC008558.1 | 2 | 539 | AAH08558.1 |

Genomic Map