Assay Details
Target Gene Details
Entrez Gene ID: | 72254 |
Gene Name: | RIKEN cDNA 1700030K09 gene |
Gene Aliases: |
- |
Location: |
Chr.8:72443880-72460541 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700030K09Rik | NM_028170.2 | 3 | 930 | NP_082446.2 |
| AK089918.1 | 3 | 871 | ||
| AK133815.1 | 3 | 934 | BAE21858.1 | |
| AK157359.1 | 2 | 141 | ||
| AK172202.1 | 2 | 139 | ||
| BC008558.1 | 3 | 884 | AAH08558.1 |

Genomic Map