Assay Details
Target Gene Details
Entrez Gene ID: | 69773 |
Gene Name: | RIKEN cDNA 1810026J23 gene |
Gene Aliases: |
- |
Location: |
Chr.9:21592722-21595970 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1810026J23Rik | NM_178619.4 | 1 | 179 | NP_848734.1 |
| AK028799.1 | BAC26126.1 | |||
| AK030602.1 | BAC27040.1 | |||
| BC057071.1 | 1 | 179 | AAH57071.1 |
Target Gene Details
Entrez Gene ID: | 74766 |
Gene Name: | Yip1 domain family, member 2 |
Gene Aliases: |
1300010K09Rik |
Location: |
Chr.9:21588682-21592831 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yipf2 | NM_001205157.1 | NP_001192086.1 | ||
| NM_001205158.1 | NP_001192087.1 | |||
| NM_138303.2 | NP_612176.1 | |||
| NR_038062.1 | ||||
| AA967152.1 |

Genomic Map