Assay Details
Target Gene Details
Entrez Gene ID: | 72254 |
Gene Name: | RIKEN cDNA 1700030K09 gene |
Gene Aliases: |
- |
Location: |
Chr.8:72443880-72460541 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700030K09Rik | NM_028170.2 | 8 | 2343 | NP_082446.2 |
| AK089918.1 | 8 | 2285 | ||
| AK133815.1 | 8 | 2347 | BAE21858.1 | |
| AK157359.1 | 7 | 1556 | ||
| AK172202.1 | 7 | 1552 | ||
| BC008558.1 | 8 | 2297 | AAH08558.1 | |
| CO043582.1 | 1 | 99 |
Target Gene Details
Entrez Gene ID: | 27967 |
Gene Name: | calcium homeostasis endoplasmic reticulum protein |
Gene Aliases: |
5730408I11Rik, D8Wsu96e, DAN16, Scaf6 |
Location: |
Chr.8:72460483-72476675 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cherp | NM_138585.3 | NP_613051.3 | ||
| AF536541.1 | AAN77182.1 | |||
| AK049765.1 | ||||
| AK144328.1 | ||||
| AK171907.1 | ||||
| BC032191.1 |
More Information
Set Membership: |
|

Genomic Map