Assay Details
Target Gene Details
Entrez Gene ID: | 269854 |
Gene Name: | N-acetyltransferase 14 |
Gene Aliases: |
BC047219 |
Location: |
Chr.7:4918888-4925006 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nat14 |
Target Gene Details
Entrez Gene ID: | 232816 |
Gene Name: | zinc finger protein 628 |
Gene Aliases: |
Zec, Znf628 |
Location: |
Chr.7:4915176-4922003 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp628 | NM_170759.2 | 3 | 1375 | NP_739565.2 |
| AF435832.1 | 1 | 1079 | AAN63612.1 | |
| AK048291.1 | 3 | 1342 | ||
| AK155214.1 | 3 | 1375 | BAE33125.1 | |
| BC056945.1 | AAH56945.1 | |||
| BC062973.1 | AAH62973.1 |
More Information
Set Membership: |
|

Genomic Map