Assay Details
Target Gene Details
Entrez Gene ID: | 74165 |
Gene Name: | F-box and leucine-rich repeat protein 22 |
Gene Aliases: |
1110004B15Rik |
Location: |
Chr.9:66508459-66514593 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxl22 | NM_175206.4 | 1 | 82 | NP_780415.1 |
| AK003404.1 | 1 | 100 | ||
| AK052175.1 | 1 | 68 | BAC34870.1 | |
| AK084845.1 | BAC39291.1 | |||
| AK085980.1 | BAC39583.1 | |||
| BC100535.1 | 1 | 78 | AAI00536.1 |

Genomic Map