Assay Details
Target Gene Details
Entrez Gene ID: | 74165 |
Gene Name: | F-box and leucine-rich repeat protein 22 |
Gene Aliases: |
1110004B15Rik |
Location: |
Chr.9:66508459-66514593 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxl22 | NM_175206.4 | 2 | 2752 | NP_780415.1 |
| AK035656.1 | 1 | 1412 | ||
| AK052175.1 | 2 | 2738 | BAC34870.1 | |
| AK084845.1 | 2 | 2668 | BAC39291.1 |

Genomic Map