Assay Details
Target Gene Details
Entrez Gene ID: | 57247 |
Gene Name: | zinc finger protein (C2H2 type) 276 |
Gene Aliases: |
AW048709, D8Ertd370e, D8Ertd377e, Znf276 |
Location: |
Chr.8:123254195-123270551 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp276 | NM_020497.2 | 1 | 481 | NP_065243.2 |
| AK028939.1 | 1 | 434 | BAC26204.1 | |
| AK043258.1 | 1 | 406 | BAC31505.1 | |
| AK133363.1 | 1 | 482 | BAE21616.1 |
More Information
Set Membership: |
|

Genomic Map