Assay Details
Target Gene Details
Entrez Gene ID: | 14087 |
Gene Name: | Fanconi anemia, complementation group A |
Gene Aliases: |
AW208693, FACA |
Location: |
Chr.8:123268244-123318651 on Build GRCm38 |
Assay Gene Location: | Within Intron 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fanca | NM_016925.3 | NP_058621.2 | ||
| AF178934.1 | AAG01633.1 | |||
| AF208116.1 | AAF63963.1 | |||
| AF208120.1 | AAF63966.1 | |||
| BC150705.1 |
Target Gene Details
Entrez Gene ID: | 57247 |
Gene Name: | zinc finger protein (C2H2 type) 276 |
Gene Aliases: |
AW048709, D8Ertd370e, D8Ertd377e, Znf276 |
Location: |
Chr.8:123254195-123270551 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp276 | NM_020497.2 | 11 | 4411 | NP_065243.2 |
| AK133363.1 | 11 | 4412 | BAE21616.1 | |
| AK163486.1 | 2 | 2402 | BAE37365.1 |
More Information
Set Membership: |
|

Genomic Map