Assay Details
Target Gene Details
Entrez Gene ID: | 94187 |
Gene Name: | zinc finger protein 423 |
Gene Aliases: |
Ebfaz, Roaz, Zfp104, Znf423, ataxia1, mKIAA0760, nur12 |
Location: |
Chr.8:87661810-87961859 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp423 | NM_033327.2 | 8 | 4334 | NP_201584.2 |
| AF188609.1 | 6 | 3703 | AAG17053.1 | |
| AK122365.1 | 8 | 4300 | BAC65647.1 | |
| AK133497.1 | 6 | 3762 | BAE21688.1 | |
| AY147407.1 | 7 | 3939 | AAN39840.1 | |
| AY256893.1 | 8 | 4233 | AAP33073.1 | |
| BB788751.1 | ||||
| BC059234.1 | 6 | 4145 | AAH59234.1 | |
| BC139028.1 | 8 | 4080 | ||
| BC139030.1 | 8 | 4080 | ||
| BU519630.1 | 5 | 557 |
More Information
Set Membership: |
|

Genomic Map