Assay Details
Target Gene Details
Entrez Gene ID: | 235040 |
Gene Name: | autophagy related 4D, cysteine peptidase |
Gene Aliases: |
9830134P12Rik, APG4-D, Apg4d, Atg4dl, Autl4 |
Location: |
Chr.9:21265285-21274837 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atg4d | NM_153583.10 | 4 | 929 | NP_705811.8 |
| AK036282.1 | 4 | 917 | ||
| AK036571.1 | 4 | 918 | BAC29484.1 | |
| AK081002.1 | 4 | 916 | BAC38110.1 | |
| AK081605.1 | 4 | 907 | BAC38269.1 | |
| AK087258.1 | 4 | 919 | ||
| AK154130.1 | 4 | 922 | BAE32395.1 | |
| BC030861.1 | 4 | 906 | AAH30861.1 | |
| BC069851.1 | 4 | 881 | AAH69851.1 |
More Information
Set Membership: |
|

Genomic Map