Assay Details
Target Gene Details
Entrez Gene ID: | 434350 |
Gene Name: | RIKEN cDNA 9430091E24 gene |
Gene Aliases: |
- |
Location: |
Chr.8:111127444-111145498 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9430091E24Rik | NR_040363.1 | |||
| NR_040364.1 | ||||
| AK020517.1 | ||||
| AK053982.1 | ||||
| BF472197.1 |
Target Gene Details
Entrez Gene ID: | 319518 |
Gene Name: | pyruvate dehydrogenase phosphatase regulatory subunit |
Gene Aliases: |
4930402E16Rik, AU018056, mKIAA1990 |
Location: |
Chr.8:111094599-111139779 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdpr | NM_198308.1 | 18 | 2636 | NP_938050.1 |
| AK029575.1 | 19 | 2806 | ||
| AK078515.1 | 12 | 1727 | ||
| AK129472.1 | 18 | 2740 | BAC98282.1 | |
| AY223867.1 | 18 | 2636 | AAP20649.1 | |
| BC120745.1 | 17 | 2566 | ||
| BC125425.1 | 17 | 2566 |
More Information
Set Membership: |
|

Genomic Map