Assay Details
Target Gene Details
Entrez Gene ID: | 67384 |
Gene Name: | BCL2-associated athanogene 4 |
Gene Aliases: |
2410112I15Rik, SODD |
Location: |
Chr.8:25764538-25785209 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bag4 | NM_026121.3 | 3 | 510 | NP_080397.1 |
| AF332863.1 | 3 | 503 | AAL99586.1 | |
| AK010765.1 | 3 | 501 | BAB27167.1 | |
| AK136899.1 | 3 | 500 | BAE23161.1 | |
| AK160316.1 | 3 | 502 | ||
| AK162658.1 | 3 | 497 | BAE37009.1 | |
| BC037239.1 | 3 | 508 | AAH37239.1 | |
| BC058518.1 | 3 | 493 | AAH58518.1 | |
| BC138201.1 | 3 | 588 | ||
| BC145986.1 | 3 | 588 |

Genomic Map