Assay Details
Target Gene Details
Entrez Gene ID: | 100042715 |
Gene Name: | predicted gene 3985 |
Gene Aliases: |
- |
Location: |
Chr.8:32888505-32950026 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm3985 | NM_001177589.1 | 4 | 2148 | NP_001171060.1 |
| AK158352.1 | 4 | 2149 | BAE34468.1 |
Target Gene Details
Entrez Gene ID: | 211323 |
Gene Name: | neuregulin 1 |
Gene Aliases: |
6030402G23Rik, ARIA, D230005F13Rik, GGF, GGFII, HRG, HRGalpha, Hgl, NDF, SMDF |
Location: |
Chr.8:31807452-32890583 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrg1 |

Genomic Map