Assay Details
Target Gene Details
Entrez Gene ID: | 244349 |
Gene Name: | K(lysine) acetyltransferase 6A |
Gene Aliases: |
1500036M03, 9930021N24Rik, MOZ, Myst3, Zfp220 |
Location: |
Chr.8:22859516-22943262 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kat6a | NM_001081149.1 | 18 | 6481 | NP_001074618.1 |
| AK054354.1 | 1 | 422 | BAC35748.1 | |
| AK148261.1 | 1 | 813 | BAE28445.1 |
More Information
Set Membership: |
|

Genomic Map