Assay Details
Target Gene Details
Entrez Gene ID: | 17960 |
Gene Name: | N-acetyl transferase 1 |
Gene Aliases: |
Nat-1 |
Location: |
Chr.8:67485170-67492582 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nat1 | NM_008673.1 | 1 | 226 | NP_032699.1 |
| BC119181.1 | ||||
| BC137747.1 | ||||
| U35885.1 | 1 | 226 | AAA78942.1 |
More Information
Set Membership: |
|

Genomic Map