Assay Details
Target Gene Details
Entrez Gene ID: | 22229 |
Gene Name: | uncoupling protein 3 (mitochondrial, proton carrier) |
Gene Aliases: |
AI645527, Slc25a9, UCP-3 |
Location: |
Chr.7:100472991-100486432 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ucp3 | NM_009464.3 | 7 | 1249 | NP_033490.1 |
| AB010742.1 | 7 | 1249 | BAA25697.1 | |
| AF032902.1 | 7 | 1216 | AAB87084.1 | |
| AF053352.1 | AAC28328.1 | |||
| BC139430.1 | 6 | 1117 | ||
| BC139431.1 | 6 | 1117 |
More Information
Set Membership: |
|

Genomic Map