Assay Details
Target Gene Details
Entrez Gene ID: | 277978 |
Gene Name: | exocyst complex component 3-like |
Gene Aliases: |
C730015A04Rik, Exoc3l1 |
Location: |
Chr.8:105289923-105296962 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exoc3l | NM_177788.4 | 5 | 978 | NP_808456.1 |
| AK050100.1 | 5 | 979 | BAC34066.1 | |
| AK140526.1 | 1 | 70 | BAE24415.1 | |
| AK172618.1 | 4 | 645 | BAE43096.1 | |
| BC094238.1 | 5 | 932 | AAH94238.1 |
More Information
Set Membership: |
|

Genomic Map