Assay Details
Target Gene Details
Entrez Gene ID: | 434350 |
Gene Name: | RIKEN cDNA 9430091E24 gene |
Gene Aliases: |
- |
Location: |
Chr.8:111127444-111145498 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9430091E24Rik | NR_040363.1 | |||
| NR_040364.1 | ||||
| AK020517.1 | ||||
| AK053982.1 | ||||
| AK080922.1 | 1 | 1510 | ||
| BF472197.1 |
More Information
Set Membership: |
|

Genomic Map