Assay Details
Target Gene Details
Entrez Gene ID: | 13036 |
Gene Name: | cathepsin H |
Gene Aliases: |
AL022844 |
Location: |
Chr.9:90054267-90076095 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsh | NM_007801.2 | 1 | 68 | NP_031827.2 |
More Information
Set Membership: |
|

Genomic Map