Assay Details
Target Gene Details
Entrez Gene ID: | 100316680 |
Gene Name: | microRNA 1903 |
Gene Aliases: |
Mirn1903, mmu-mir-1903 |
Location: |
Chr.8:128359241-128359320 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir1903 | NR_035436.1 |
Target Gene Details
Entrez Gene ID: | 18186 |
Gene Name: | neuropilin 1 |
Gene Aliases: |
C530029I03, NP-1, NPN-1, Npn1, Nrp |
Location: |
Chr.8:128358285-128505476 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrp1 | NM_008737.2 | 1 | 281 | NP_032763.2 |
| AK030358.1 | 1 | 94 | ||
| AK040447.1 | 1 | 102 | ||
| AK082992.1 | 1 | 334 | ||
| AK083052.1 | 1 | 308 | ||
| AK135428.1 | ||||
| AK141964.1 | 1 | 72 | BAE24901.1 | |
| AK148372.1 | 1 | 257 | BAE28513.1 | |
| AK159219.1 | 1 | 72 | ||
| AK164378.1 | 1 | 283 | ||
| AK171933.1 | 2 | 382 | ||
| BC051447.1 | 1 | 85 | AAH51447.1 | |
| BC060129.1 | 1 | 61 | AAH60129.1 | |
| D50086.1 | 1 | 141 | BAA08789.1 |
More Information
Set Membership: |
|

Genomic Map