Assay Details
Target Gene Details
Entrez Gene ID: | 69165 |
Gene Name: | CD209b antigen |
Gene Aliases: |
1810030I22Rik, DC-SIGNR1, OtB7, SIGNR1, mSIGNR1 |
Location: |
Chr.8:3917165-3926841 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd209b | NM_001037800.3 | 9 | 1561 | NP_001032889.3 |
| NM_001287211.1 | 10 | 1624 | NP_001274140.1 | |
| NM_026972.5 | 10 | 1651 | NP_081248.4 | |
| NR_104430.2 | 9 | 1499 | ||
| AF424802.1 | 4 | 1054 | AAN75597.1 | |
| AF424803.1 | 9 | 1507 | AAN75598.1 | |
| AK138999.1 | 10 | 1625 | BAE23855.1 |
More Information
Set Membership: |
|

Genomic Map