Assay Details
Target Gene Details
Entrez Gene ID: | 69142 |
Gene Name: | CD209f antigen |
Gene Aliases: |
1810029C22Rik, AI647041, SIGNR8 |
Location: |
Chr.8:4102784-4105838 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd209f | NM_026956.2 | 8 | 722 | NP_081232.2 |
| AK007638.1 | 8 | 779 | ||
| BC021323.1 | 8 | 803 | ||
| BC038121.1 | 8 | 1125 | AAH38121.1 | |
| BC046292.1 | 6 | 763 | AAH46292.1 | |
| BC052782.1 | 8 | 713 | AAH52782.1 | |
| BC099497.1 | 8 | 717 | AAH99497.1 |
More Information
Set Membership: |
|

Genomic Map