Assay Details
Target Gene Details
Entrez Gene ID: | 14232 |
Gene Name: | FK506 binding protein 8 |
Gene Aliases: |
38kDa, FKBP-38, FKBP-8, Fkbp38 |
Location: |
Chr.8:70527743-70535328 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp8 | NM_001111066.1 | 2 | 191 | NP_001104536.1 |
| NM_001199631.1 | 2 | 287 | NP_001186560.1 | |
| NM_010223.2 | 2 | 191 | NP_034353.2 | |
| AK040736.1 | 2 | 124 | ||
| AK088739.1 | 2 | 126 | BAC40541.1 | |
| AK146122.1 | 2 | 186 | BAE26916.1 | |
| AK152198.1 | 2 | 188 | BAE31027.1 | |
| AK153000.1 | 2 | 184 | ||
| AK158155.1 | 2 | 174 | ||
| AK165281.1 | 2 | 184 | BAE38118.1 | |
| AK167663.1 | 2 | 174 | BAE39713.1 | |
| AK170993.1 | 1 | 1745 | ||
| AK190318.1 | 1 | 102 | ||
| AK197498.1 | 1 | 102 | ||
| AK211059.1 | 1 | 102 | ||
| AK216136.1 | ||||
| AY225340.1 | 1 | 74 | AAO39021.1 | |
| AY278608.1 | 2 | 150 | AAQ84562.1 | |
| BB842097.1 | 2 | 191 | ||
| BC003739.1 | 1 | 107 | AAH03739.1 | |
| BC027808.1 | 2 | 141 | AAH27808.1 |
More Information
Set Membership: |
|

Genomic Map