Assay Details
Target Gene Details
Entrez Gene ID: | 66234 |
Gene Name: | methylsterol monoxygenase 1 |
Gene Aliases: |
1500001G16Rik, C78600, DESP4, ERG25, Sc4mol |
Location: |
Chr.8:64718145-64733578 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msmo1 | NM_025436.2 | 6 | 1657 | NP_079712.1 |
| AK005090.1 | 6 | 1658 | BAB23811.1 | |
| AK005441.1 | 6 | 1652 | BAB24035.1 | |
| AK010570.1 | 4 | 1226 | ||
| AK045059.1 | 6 | 1628 | BAC32201.1 | |
| AK146684.1 | 6 | 1626 | BAE27358.1 | |
| AK151135.1 | 6 | 1658 | BAE30143.1 | |
| AK154682.1 | 5 | 2682 | ||
| AK169017.1 | 6 | 1627 | BAE40814.1 | |
| BC006802.1 | 6 | 1630 | AAH06802.1 |
More Information
Set Membership: |
|

Genomic Map