Assay Details
Target Gene Details
Entrez Gene ID: | 12556 |
Gene Name: | cadherin 16 |
Gene Aliases: |
- |
Location: |
Chr.8:104601915-104624396 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh16 | NM_001252627.1 | 17 | 2738 | NP_001239556.1 |
| NM_001252628.1 | NP_001239557.1 | |||
| NM_007663.3 | 18 | 2828 | NP_031689.1 | |
| AF016271.1 | 18 | 2696 | AAC34254.1 | |
| AJ609635.1 | 18 | 2695 | CAE82011.1 | |
| AK052636.1 | BAC35072.1 | |||
| AK093905.1 | 18 | 2626 | ||
| AK164546.1 | 17 | 2739 | BAE37833.1 | |
| BC015251.1 | 18 | 2676 | AAH15251.1 |
More Information
Set Membership: |
|

Genomic Map