Assay Details
Target Gene Details
Entrez Gene ID: | 72330 |
Gene Name: | kelch-like 40 |
Gene Aliases: |
2310024D23Rik, Kbtbd5 |
Location: |
Chr.9:121777101-121791002 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl40 | NM_028202.3 | 6 | 2004 | NP_082478.1 |
| AK009491.1 | 6 | 1920 | BAB26321.1 | |
| AK078669.1 | 6 | 1918 | ||
| AK160799.1 | 6 | 1864 | BAE36021.1 |
More Information
Set Membership: |
|

Genomic Map