Assay Details
Target Gene Details
Entrez Gene ID: | 208098 |
Gene Name: | pannexin 3 |
Gene Aliases: |
3230401P04, 4833413G11Rik |
Location: |
Chr.9:37659902-37669222 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Panx3 | NM_172454.2 | 1 | 116 | NP_766042.2 |
| AK014690.1 | 1 | 85 | ||
| AK028288.1 | 1 | 83 | BAC25860.1 | |
| AK036933.1 | 1 | 72 | BAC29644.1 | |
| BB849008.1 | 1 | 130 | ||
| BC052822.1 | 1 | 84 | AAH52822.1 |

Genomic Map