Assay Details
Target Gene Details
Entrez Gene ID: | 327826 |
Gene Name: | fibroblast growth factor receptor substrate 2 |
Gene Aliases: |
4732458E18, C330018A15Rik, Frs2alpha, SNT1 |
Location: |
Chr.10:117070127-117148474 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Frs2 | NM_177798.3 | 10 | 1726 | NP_808466.1 |
| AK028813.1 | 10 | 1742 | BAC26132.1 | |
| BC043109.1 | 10 | 1726 | AAH43109.1 | |
| BC055334.1 | 10 | 1762 | AAH55334.1 |

Genomic Map