Assay Details
Target Gene Details
Entrez Gene ID: | 78670 |
Gene Name: | pleckstrin homology domain containing, family J member 1 |
Gene Aliases: |
9530063M10Rik |
Location: |
Chr.10:80796099-80798626 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekhj1 | NM_023900.2 | 1 | 266 | NP_076389.2 |
| AK088567.1 | 1 | 227 | ||
| DV657752.1 | 1 | 266 |
More Information
Set Membership: |
|

Genomic Map