Assay Details
Target Gene Details
Entrez Gene ID: | 110312 |
Gene Name: | pro-melanin-concentrating hormone |
Gene Aliases: |
A230109K23Rik |
Location: |
Chr.10:88091072-88092374 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmch | NM_029971.2 | 1 | 100 | NP_084247.1 |
| AK020723.1 | 1 | 100 | BAB32189.1 | |
| AK138363.1 | 1 | 103 | BAE23633.1 | |
| BC048534.1 | 1 | 100 | AAH48534.1 |
More Information
Set Membership: |
|

Genomic Map