Assay Details
Target Gene Details
Entrez Gene ID: | 244694 |
Gene Name: | lysine (K)-specific demethylase 4D |
Gene Aliases: |
4932416A15, Jmjd2d |
Location: |
Chr.9:14462548-14500522 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm4d | NM_173433.2 | 1 | 123 | NP_775609.2 |
| AK030020.1 | 1 | 123 | BAC26740.1 | |
| AK155227.1 | BAE33135.1 |
More Information
Set Membership: |
|

Genomic Map