Assay Details
Target Gene Details
Entrez Gene ID: | 268301 |
Gene Name: | sosondowah ankyrin repeat domain family member C |
Gene Aliases: |
4921515A04Rik, 9430099J10Rik, Ankrd57, C820004L04Rik |
Location: |
Chr.10:59221922-59226434 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sowahc | NM_172939.3 | 1 | 1500 | NP_766527.3 |
AK030944.1 | 1 | 1501 | BAC27191.1 | |
AK050497.1 | 1 | 1470 | BAC34291.1 |
