Assay Details
Target Gene Details
Entrez Gene ID: | 170483 |
Gene Name: | glutamate receptor, ionotropic, NMDA3B |
Gene Aliases: |
GluN3B, NR3B |
Location: |
Chr.10:79970397-79977190 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grin3b | NM_130455.2 | 3 | 2050 | NP_569722.1 |
| AF373861.1 | 1 | 799 | AAL40418.1 | |
| AF396649.1 | 3 | 2059 | AAL10018.1 | |
| AY062435.1 | 3 | 2050 | AAL38983.1 | |
| BC115747.1 | 1 | 137 |
More Information
Set Membership: |
|

Genomic Map