Assay Details
Target Gene Details
Entrez Gene ID: | 170483 |
Gene Name: | glutamate receptor, ionotropic, NMDA3B |
Gene Aliases: |
GluN3B, NR3B |
Location: |
Chr.10:79970397-79977190 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grin3b | NM_130455.2 | 9 | 3203 | NP_569722.1 |
| AF396649.1 | 9 | 3212 | AAL10018.1 | |
| AY062435.1 | 9 | 3203 | AAL38983.1 | |
| BC115747.1 |
Target Gene Details
Entrez Gene ID: | 216157 |
Gene Name: | transmembrane protein 259 |
Gene Aliases: |
AC087114.7gm2, ORF61, R32184_3, mFLJ00277 |
Location: |
Chr.10:79977120-79984330 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem259 | NM_001003949.3 | NP_001003949.2 | ||
| BC005494.1 | AAH05494.1 | |||
| BC052787.1 | AAH52787.1 | |||
| BC100297.1 | AAI00298.1 |
More Information
Set Membership: |
|

Genomic Map