Assay Details
Target Gene Details
Entrez Gene ID: | 12974 |
Gene Name: | citrate synthase |
Gene Aliases: |
2610511A05Rik, 9030605P22Rik, Ahl4, BB234005, Cis |
Location: |
Chr.10:128337832-128362479 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cs | NM_026444.3 | 11 | 2319 | NP_080720.1 |
| AK012104.1 | 5 | 1497 | ||
| AK149990.1 | 11 | 2418 | BAE29218.1 | |
| AK163273.1 | 11 | 2344 | BAE37268.1 | |
| BC029754.1 | 11 | 2319 | AAH29754.1 |
More Information
Set Membership: |
|

Genomic Map