Assay Details
Target Gene Details
Entrez Gene ID: | 14609 |
Gene Name: | gap junction protein, alpha 1 |
Gene Aliases: |
AU042049, AW546267, Cnx43, Cx43, Cx43alpha1, Gja-1, Npm1, connexin43 |
Location: |
Chr.10:56377201-56390423 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gja1 | NM_010288.3 | 2 | 1184 | NP_034418.1 |
| AK029291.1 | 2 | 1041 | BAC26375.1 | |
| AK036979.1 | 2 | 1155 | BAC29656.1 | |
| AK081668.1 | 1 | 131 | BAC38285.1 | |
| AK145692.1 | 2 | 1154 | BAE26592.1 | |
| AK165986.1 | 2 | 1041 | BAE38502.1 | |
| AK171955.1 | 2 | 1153 | ||
| BC006894.1 | 2 | 1137 | AAH06894.1 | |
| BC055375.1 | 2 | 1107 | AAH55375.1 | |
| CT010327.1 | 1 | 952 | CAJ18535.1 | |
| M63801.1 | 2 | 1172 | AAA53027.1 | |
| X61576.1 | 2 | 1038 | CAA43778.1 |
More Information
Set Membership: |
|

Genomic Map