Assay Details
Target Gene Details
Entrez Gene ID: | 26396 |
Gene Name: | mitogen-activated protein kinase kinase 2 |
Gene Aliases: |
AA589381, MEK2, MK2, Prkmk2 |
Location: |
Chr.10:81105912-81124697 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map2k2 | NM_023138.4 | 11 | 1838 | NP_075627.2 |
| AK009392.1 | 11 | 1855 | BAB26261.1 | |
| AK036822.1 | 3 | 664 | BAC29592.1 | |
| AK140101.1 | 1 | 513 | BAE24237.1 | |
| AK192399.1 | ||||
| AK212483.1 | 1 | 48 | ||
| AK220136.1 |
More Information
Set Membership: |
|

Genomic Map