Assay Details
Target Gene Details
Entrez Gene ID: | 235040 |
Gene Name: | autophagy related 4D, cysteine peptidase |
Gene Aliases: |
9830134P12Rik, APG4-D, Apg4d, Atg4dl, Autl4 |
Location: |
Chr.9:21265285-21274837 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atg4d | NM_153583.10 | 10 | 1910 | NP_705811.8 |
| AK036282.1 | 10 | 1898 | ||
| AK036571.1 | 10 | 1899 | BAC29484.1 | |
| AK081605.1 | BAC38269.1 | |||
| AK087258.1 | ||||
| AK154130.1 | BAE32395.1 | |||
| AK213738.1 | ||||
| AK215761.1 | ||||
| BC030861.1 | AAH30861.1 | |||
| BC069851.1 | AAH69851.1 |
Target Gene Details
Entrez Gene ID: | 215194 |
Gene Name: | KRI1 homolog (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.9:21273457-21287969 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kri1 | NM_145416.3 | 19 | 2332 | NP_663391.2 |
| BC021438.1 | 19 | 2322 | AAH21438.1 | |
| BQ557175.1 | 1 | 262 | ||
| CK795617.1 | 1 | 366 |
More Information
Set Membership: |
|

Genomic Map