Assay Details
Target Gene Details
Entrez Gene ID: | 67933 |
Gene Name: | host cell factor C2 |
Gene Aliases: |
1700129L13Rik, AU019838 |
Location: |
Chr.10:82697497-82795865 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hcfc2 | NM_001081218.1 | 14 | 2079 | NP_001074687.1 |
| AK007317.1 | 5 | 562 | BAB24953.1 | |
| BC116411.1 | 7 | 871 | ||
| BC127176.1 | 7 | 871 |

Genomic Map