Assay Details
Target Gene Details
Entrez Gene ID: | 66925 |
Gene Name: | succinate dehydrogenase complex, subunit D, integral membrane protein |
Gene Aliases: |
3110001M13Rik, AVLL5809, C78570, PRO19626 |
Location: |
Chr.9:50596340-50603849 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdhd | NM_025848.3 | 4 | 533 | NP_080124.1 |
| AK013503.1 | 4 | 498 | BAB28885.1 | |
| AK013962.2 | 4 | 501 | BAB29086.2 | |
| AK135970.1 | 4 | 493 | BAE22752.1 | |
| BC145731.1 | 4 | 493 | ||
| BC145733.1 | 4 | 493 |
More Information
Set Membership: |
|

Genomic Map