Assay Details
Target Gene Details
Entrez Gene ID: | 20347 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3B |
Gene Aliases: |
SemA, Semaa, sema5, semaV |
Location: |
Chr.9:107597674-107609303 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema3b | NM_001042779.2 | 20 | 2946 | NP_001036244.1 |
| NM_001291537.1 | 20 | 3210 | NP_001278466.1 | |
| NM_001291538.1 | 20 | 3204 | NP_001278467.1 | |
| NM_001291539.1 | 19 | 2887 | NP_001278468.1 | |
| NM_009153.3 | 18 | 2756 | NP_033179.2 | |
| NR_111986.1 | 18 | 2699 | ||
| AK014333.1 | 18 | 2635 | ||
| AK033597.1 | 19 | 2897 | ||
| AK037034.1 | 20 | 2935 | ||
| AK043634.1 | 19 | 2840 | ||
| AK048364.1 | 19 | 3168 | ||
| AK148232.1 | 20 | 2907 | ||
| AK180933.1 | 1 | 52 | ||
| AK182150.1 | 1 | 52 | ||
| AK218182.1 | 1 | 318 | ||
| BB787270.1 | 1 | 92 | ||
| BC090669.1 | 19 | 2867 | ||
| X85990.1 | 18 | 2753 | CAA59982.1 |
More Information
Set Membership: |
|

Genomic Map