Assay Details
Target Gene Details
Entrez Gene ID: | 56395 |
Gene Name: | transmembrane protein 115 |
Gene Aliases: |
C78915, Pl6, Pp6 |
Location: |
Chr.9:107533945-107538656 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem115 | NM_019704.2 | 2 | 1779 | NP_062678.1 |
| AF134238.1 | 2 | 1779 | AAD24193.1 | |
| AK043834.1 | 2 | 1707 | BAC31672.1 | |
| AK154202.1 | 2 | 1704 | BAE32435.1 | |
| AK164772.1 | 1 | 85 | BAE37911.1 | |
| AK170756.1 | 2 | 1638 | BAE42007.1 | |
| AK209295.1 | 1 | 178 | ||
| BC009099.1 | 2 | 623 | AAH09099.1 | |
| BC019473.1 | 2 | 1635 | AAH19473.1 |
More Information
Set Membership: |
|

Genomic Map