Assay Details
Target Gene Details
Entrez Gene ID: | 103268 |
Gene Name: | centrosomal protein 57-like 1 |
Gene Aliases: |
2410017P07Rik, 4930484D11Rik, AI607327, cep57R |
Location: |
Chr.10:41718840-41809868 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep57l1 | NM_001243074.1 | 11 | 1698 | NP_001230003.1 |
| NM_001243075.1 | 10 | 1476 | NP_001230004.1 | |
| NM_026643.5 | 11 | 1616 | NP_080919.3 | |
| NM_029132.2 | 12 | 1771 | NP_083408.1 | |
| AF397908.1 | 12 | 1699 | ||
| AK010530.1 | 12 | 1722 | ||
| AK012710.1 | 10 | 1422 | BAB28426.1 | |
| BC021375.1 | 11 | 1708 | AAH21375.1 | |
| BC138230.1 | 11 | 1619 | ||
| BM245951.2 | 1 | 536 |

Genomic Map